Learn More
Med Vet International Miltex Vantage Crile Hemostatic Forceps, 5.5in, Curved
Catalog No. 502522261
Encompass_Preferred
The Miltex Vantage V97-44 Crile forceps are made from high quality Japanese stainless steel and measure 5.5" in length. They have a finger ring handle with a ratchet lock and curved tips with serrations.
Catalog No. 50-252-2261 Supplier Med Vet International Supplier No. MILV9744
The Miltex Vantage V97-44 Crile forceps are made from high quality Japanese stainless steel and measure 5.5" in length. They have a finger ring handle with a ratchet lock and curved tips with serrations.