Learn More
Med Vet International Miltex Crile Hemostatic Forceps, 5.5in., Straight
Catalog No. 502828991
Encompass_Preferred
The Miltex 7-42 Crile forceps are made of OR grade German stainless steel and measure 5.5" in length. They have a finger ring handle with ratchet lock and straight tips with serrations.
Catalog No. 50-282-8991 Supplier Med Vet International Supplier No. MIL742
The Miltex 7-42 Crile forceps are made of OR grade German stainless steel and measure 5.5" in length. They have a finger ring handle with ratchet lock and straight tips with serrations.