Learn More
Med Vet International Miltex Meisterhand Crile Forceps, 6.25in, Curved, German, Mh7-48
Supplier:  Med Vet International MILMH748
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Crile Forceps. Crile Forceps are one of the most commonly used hemostatic forceps. They are designed for clamping blood vessels, small arteries and tissue to control bleeding They are made from premium-grade German stainless steel. Specifications: 6.25in, Curved, MH7-48
Catalog No. 50-283-0963