Learn More
Med Vet International Miltex Crile Hemostatic Forceps, 6.25in., Straight
Catalog No. 502828992
Encompass_Preferred
The Miltex 7-46 Crile forceps are made of OR grade German stainless steel and measure 6.25" in length. They have a finger ring handle with a ratchet lock and straight tips with serrations.
Catalog No. 50-282-8992 Supplier Med Vet International Supplier No. MIL746
The Miltex 7-46 Crile forceps are made of OR grade German stainless steel and measure 6.25" in length. They have a finger ring handle with a ratchet lock and straight tips with serrations.