Learn More
Med Vet International Crile Forceps, 6.25", Curved, German Grade, Von Klaus
Catalog No. 503046053
Encompass_Preferred
6.25in. Crile Forceps is a German grade surgical instrument. These forceps are curved and are commonly used for clamping blood vessels or tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6053 Supplier Med Vet International Supplier No. VK1157316
6.25in. Crile Forceps is a German grade surgical instrument. These forceps are curved and are commonly used for clamping blood vessels or tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.