Learn More
Med Vet International Crile Forceps, 5.5-in Curved, Von Klaus German Surgical Steel
Catalog No. 503046613
Encompass_Preferred
These forceps are curved and are commonly used for clamping blood vessels or tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6613 Supplier Med Vet International Supplier No. VK1157314
These forceps are curved and are commonly used for clamping blood vessels or tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.