Learn More
BURKLE INC Forceps, disposable, pointed/angled, 5.12 in. (130mm), PS
Catalog No. 503081194
Encompass_Preferred
The disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.
The fluted handles ensure optimum and secure handling as well as precise work.
The light plastic forceps for single use allow for fatigue-free work. The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined.
Catalog No. 50-308-1194 Supplier BURKLE INC Supplier No. 53780040
The disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.
The fluted handles ensure optimum and secure handling as well as precise work.
The light plastic forceps for single use allow for fatigue-free work. The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined.