Learn More
Med Vet International Miltex Vantage Doyen Intestinal Hemostatic Forceps, 9in, Straight, Longitudinal Serrations
Catalog No. 502522282
Encompass_Preferred
The Miltex Vantage V916-160 Doyen intestinal forceps are made from high quality Japanese stainless steel and measure 9" in length. They have a finger ring handle with a ratchet lock and straight, flexible blades with longitudinal serrations
Catalog No. 50-252-2282 Supplier Med Vet International Supplier No. MILV916160
The Miltex Vantage V916-160 Doyen intestinal forceps are made from high quality Japanese stainless steel and measure 9" in length. They have a finger ring handle with a ratchet lock and straight, flexible blades with longitudinal serrations