Learn More
Med Vet International 4.75-in Adson Thumb Dressing Forceps, Standard Pattern Dressing, Serrated, Von Klas German Surgical Steel
Catalog No. 502522149
Encompass_Preferred
This 4.75-in Adson Thumb forceps is a German surgical steel instrument. It's used for holding and manipulating delicate tissue. It has a standard pattern and is serrated. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-252-2149 Supplier Med Vet International Supplier No. VK1062012
This 4.75-in Adson Thumb forceps is a German surgical steel instrument. It's used for holding and manipulating delicate tissue. It has a standard pattern and is serrated. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.