Learn More
Med Vet International Miltex MeisterHand Adson Dressing Forceps, 4.75in, Delicate, Serrated, German, MH6-118
Catalog No. 502522288
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Adson Dressing Forceps. Adson Dressing (Thumb) Forceps are used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They are made from premium-grade German stainless steel. Specifications: 4.75in, Delicate, Serrated, MH6-118
Catalog No. 50-252-2288 Supplier Med Vet International Supplier No. MILMH6118
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Adson Dressing Forceps. Adson Dressing (Thumb) Forceps are used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They are made from premium-grade German stainless steel. Specifications: 4.75in, Delicate, Serrated, MH6-118