Learn More
Fine Science Tools Dumont #7 Forceps, Curved, Dumoxel, 0.07 x 0.04 mm, 11.5 cm
SDP
Catalog No. NC9457303
Encompass
Style: #7/ Tip Shape: Curved/ Tips: Biologie/ Tip Dimensions: 0.07 x 0.04mm/ Alloy / Material: Dumoxel/ Length: 11.5cm
Catalog No. NC9457303 Supplier Fine Science Tools Supplier No. 1127230
Style: #7/ Tip Shape: Curved/ Tips: Biologie/ Tip Dimensions: 0.07 x 0.04mm/ Alloy / Material: Dumoxel/ Length: 11.5cm