Learn More
Fine Science Tools Dumont #7 Forceps, Curved, Dumostar, 0.07 x 0.04 mm, 11.5 cm
SDP
Supplier:  Fine Science Tools 1129710
Encompass
Style: #7/ Tip Shape: Curved/ Tips: Biologie/ Tip Dimensions: 0.07 x 0.04mm/ Alloy / Material: Dumostar/ Length: 11.5cm
Catalog No. NC9781697