Learn More
TWD Scientific LLC Blunt Tip Disposable Forceps, Sterile, Endotoxin-Free, Blue. 100/Pack, 12 Packs/Case
SDP
Catalog No. NC2380034 Shop All TWD Scientific LLC Products
Encompass_Preferred
Serrated blunt tip disposable forceps, polystyrene, sterile, endotoxin-free, blue. Length: 4.9". 100/Pack, 12 Packs/Case
Catalog No. NC2380034 Supplier TWD Scientific LLC Supplier No. DF8088SENDOCS
Serrated blunt tip disposable forceps, polystyrene, sterile, endotoxin-free, blue. Length: 4.9". 100/Pack, 12 Packs/Case