Learn More
Med Vet International Miltex Vantage Extracting Forceps, Size 53L, V90-53L
Catalog No. 502830408
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Vantage Extracting Forceps. Extraction forceps are used along with elevators to extract teeth. Each extraction forcep is designed for a particular area of the mouth. The beaks are designed to fit around the cervical portion of the tooth. They are made from premium-grade surgical stainless steel. Specifications: Size 53L, V90-53L
Catalog No. 50-283-0408 Supplier Med Vet International Supplier No. MILV9053L
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Vantage Extracting Forceps. Extraction forceps are used along with elevators to extract teeth. Each extraction forcep is designed for a particular area of the mouth. The beaks are designed to fit around the cervical portion of the tooth. They are made from premium-grade surgical stainless steel. Specifications: Size 53L, V90-53L