Learn More
Med Vet International Miltex Eye Dressing Forceps, 4in, Half Curved, Standard Pattern, 0.8mm Wide Serrated Tips
Catalog No. 502643553
Encompass_Preferred
The Miltex 18-782 eye dressing forceps are made of OR grade German stainless steel and measure 4" in length. They have a non-locking thumb handle and half curved, 0.8mm wide serrated tips.
Catalog No. 50-264-3553 Supplier Med Vet International Supplier No. MIL18782
The Miltex 18-782 eye dressing forceps are made of OR grade German stainless steel and measure 4" in length. They have a non-locking thumb handle and half curved, 0.8mm wide serrated tips.