Learn More
Med Vet International Miltex Vantage Eye Dressing Forceps, 4in, Half Curved, Standard Pattern, 0.8mm Wide Serrated Tips
Catalog No. 502522278
Encompass_Preferred
The Miltex Vantage V918-782 eye dressing forceps are made from high quality Japanese stainless steel and measure 4" in length. They have a non-locking thumb handle and half curved, 0.8mm wide serrated tips.
Catalog No. 50-252-2278 Supplier Med Vet International Supplier No. MILV918782
The Miltex Vantage V918-782 eye dressing forceps are made from high quality Japanese stainless steel and measure 4" in length. They have a non-locking thumb handle and half curved, 0.8mm wide serrated tips.