Learn More
World Precision Instrument Gillies Dissecting Forceps, 15.5cm, Straight, 1x2 teeth
SDP
Catalog No. 50822625
Encompass
Gillies Dissecting Forceps, also known as Gillies forceps or Gillies skin hooks, are often used in plastic and reconstructive surgery, and are perfect for the fine work of microdissection. Gillies forceps have a 1x2 tooth configuration for a secure grip. The tips are narrow, pointed, and serrated, allowing you deeper access into incisions. The serrated and toothed tips, providing a secure grip on delicate tissues without causing excessive trauma or damage. The narrow design of the Gillies forceps tips allow for precise dissection and manipulation of small structures, such as blood vessels, nerves, and delicate skin flaps. They come as standard or with tungsten carbide inserts. Tungsten carbide (TC) is extremely hard and durable, giving you a long-lasting pair of forceps.
  • 15.5 cm long
  • Straight
  • 1 x 2 Teeth
Catalog No. 50-822-625 Supplier World Precision Instrument Supplier No. 501266
Gillies Dissecting Forceps, also known as Gillies forceps or Gillies skin hooks, are often used in plastic and reconstructive surgery, and are perfect for the fine work of microdissection. Gillies forceps have a 1x2 tooth configuration for a secure grip. The tips are narrow, pointed, and serrated, allowing you deeper access into incisions. The serrated and toothed tips, providing a secure grip on delicate tissues without causing excessive trauma or damage. The narrow design of the Gillies forceps tips allow for precise dissection and manipulation of small structures, such as blood vessels, nerves, and delicate skin flaps. They come as standard or with tungsten carbide inserts. Tungsten carbide (TC) is extremely hard and durable, giving you a long-lasting pair of forceps.
  • 15.5 cm long
  • Straight
  • 1 x 2 Teeth