Learn More
World Precision Instrument Gillies Dissecting Forceps, 15.5cm, Straight, 1x2 teeth, German
SDP
Supplier:  World Precision Instrument 501266G
Encompass
Gillies Dissecting Forceps, also known as Gillies forceps or Gillies skin hooks, are often used in plastic and reconstructive surgery, and are perfect for the fine work of microdissection. Gillies forceps have a 1x2 tooth configuration for a secure grip. The tips are narrow, pointed, and serrated, allowing you deeper access into incisions. The serrated and toothed tips, providing a secure grip on delicate tissues without causing excessive trauma or damage. The narrow design of the Gillies forceps tips allow for precise dissection and manipulation of small structures, such as blood vessels, nerves, and delicate skin flaps. They come as standard or with tungsten carbide inserts. Tungsten carbide (TC) is extremely hard and durable, giving you a long-lasting pair of forceps. With a high coefficient of friction, TC forceps offer enhanced gripping ability for a secure hold with minimal friction against tissues. The reduced friction means less tissue damage as a result of handling.
Catalog No. 50-822-626
 Restricted Item