Learn More
World Precision Instrument Graefe Forceps, 7cm, Curved, 0.7mm 1x2 Teeth
SDP
Supplier:  World Precision Instrument 14141
Encompass
Graefe Forceps, also known as Graefe's cataract forceps, are a surgical instrument commonly used in ophthalmic surgery or other microsurgeries handling delicate tissues. Named after Albrecht von Graefe, a German ophthalmologist who developed the instrument in the 1800s, they are typically used to remove cataracts from the eye, but may also be used for foreign body removal from the eye or repairing corneal incisions. The forceps may be curved or straight. A curved shape ensures more maneuverability inside the eye without causing damage to surrounding tissues. Straight Graefe forceps can be used to remove foreign objects from the eye, to manipulate the cornea during procedures such as corneal grafts, to grasp and manipulate delicate tissues during glaucoma surgery, or to manipulate or remove small pieces of tissue or membranes during retinal surgery. Our Graefe Forceps have a fine (0.7mm) tip with 1x2 teeth.
Catalog No. 50-822-263