Learn More
Med Vet International Miltex Vantage Graefe Fixation Forceps, 4.375in, Fine Teeth
Catalog No. 502522277
Encompass_Preferred
The Miltex Vantage V918-850 Graefe fixation forceps are made from high quality Japanese stainless steel and measure 4.375" in length. They have a non-locking thumb handle and straight tips with fine teeth.
Catalog No. 50-252-2277 Supplier Med Vet International Supplier No. MILV918850
The Miltex Vantage V918-850 Graefe fixation forceps are made from high quality Japanese stainless steel and measure 4.375" in length. They have a non-locking thumb handle and straight tips with fine teeth.