Learn More
Med Vet International Miltex Vantage Halsted Mosquito Hemostatic Forceps, 5in, Curved
Catalog No. 502522262
Encompass_Preferred
The Miltex Vantage V97-4 Halsted mosquito forceps are made from high quality Japanese stainless steel and measure 5" in length. They have a finger ring handle with a ratchet lock and curved tips that are serrated.
Catalog No. 50-252-2262 Supplier Med Vet International Supplier No. MILV974
The Miltex Vantage V97-4 Halsted mosquito forceps are made from high quality Japanese stainless steel and measure 5" in length. They have a finger ring handle with a ratchet lock and curved tips that are serrated.