Learn More
Med Vet International Rochster-Carmalt Forceps, 6 1/4in, Curved, German Grade, Von Klas
Catalog No. 502522148
Encompass_Preferred
This 6 1/4in. Rochester-Carmalt forceps is a German grade surgical instrument. It is curved with longitudinal serrations and cross-serrated tips. Rochester-Carmalt forceps are a type of hemostatic clamp that is used to stop blood flow of large vessels or to grip tough tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-252-2148 Supplier Med Vet International Supplier No. VK1163716
This 6 1/4in. Rochester-Carmalt forceps is a German grade surgical instrument. It is curved with longitudinal serrations and cross-serrated tips. Rochester-Carmalt forceps are a type of hemostatic clamp that is used to stop blood flow of large vessels or to grip tough tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.