Learn More
World Precision Instrument Microintraocular Forceps, 14cm, 3 x 0.6 x 0.4mm Jaws
SDP
Catalog No. 50822541
Encompass
Microintraocular Forceps, also known as microforceps or microsurgical forceps, are specialized surgical instruments used in ophthalmic procedures, particularly in intraocular surgeries. They have a fine, slender profile and fine tips that allow for precise grasping, manipulation, and dissection of tissues within the eye. They may also be used to manipulate fine sutures, remove fragments or foreign bodies from the eye, or perform intricate tissue dissection. These intraocular forceps have straight tips. You can choose from the smaller set (tip size: 2.5x0.3x0.25mm LWH) or the larger set (tip size 3.0x0.6x0.4mm LWH). The smaller set has a maximum opening of 1.5mm and the larger set has a maximum opening of 1.4mm. These are made of surgical grade stainless steel, and they are autoclavable.
Catalog No. 50-822-541 Supplier World Precision Instrument Supplier No. 501212
Microintraocular Forceps, also known as microforceps or microsurgical forceps, are specialized surgical instruments used in ophthalmic procedures, particularly in intraocular surgeries. They have a fine, slender profile and fine tips that allow for precise grasping, manipulation, and dissection of tissues within the eye. They may also be used to manipulate fine sutures, remove fragments or foreign bodies from the eye, or perform intricate tissue dissection. These intraocular forceps have straight tips. You can choose from the smaller set (tip size: 2.5x0.3x0.25mm LWH) or the larger set (tip size 3.0x0.6x0.4mm LWH). The smaller set has a maximum opening of 1.5mm and the larger set has a maximum opening of 1.4mm. These are made of surgical grade stainless steel, and they are autoclavable.