Learn More
Med Vet International Bishop-Harmon Tissue Forceps, 3.375, 1x2 Teeth, Straight, German Grade, Von Klaus
Catalog No. 503046063
Encompass_Preferred
3.375in. Bishop-Harmon tissue forceps is a German grade surgical instrument generally used in ophthalmic procedures. They are delicate forceps with 1x2 teeth and they are straight. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6063 Supplier Med Vet International Supplier No. VK1601522
3.375in. Bishop-Harmon tissue forceps is a German grade surgical instrument generally used in ophthalmic procedures. They are delicate forceps with 1x2 teeth and they are straight. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.