Learn More
Fine Science Tools Iris Forceps, Stainless Steel, Straight, 7 cm
SDP
Catalog No. NC9615154
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.6 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 7cm
Catalog No. NC9615154 Supplier Fine Science Tools Supplier No. 1106407
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.6 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 7cm