Learn More
World Precision Instrument Kelly Forceps, 14cm, Curved, German
SDP
Catalog No. 50822648
Encompass
Kelly hemostatic forceps are used to clamp blood vessels and hold heavy tissues in place. Kelly forceps, both straight and curved, are available. Our forceps feature a locking ratchet and wider finger rings for easy handling.
Catalog No. 50-822-648 Supplier World Precision Instrument Supplier No. 501288G
Kelly hemostatic forceps are used to clamp blood vessels and hold heavy tissues in place. Kelly forceps, both straight and curved, are available. Our forceps feature a locking ratchet and wider finger rings for easy handling.