Learn More
World Precision Instrument Kelly Hemostatic Forceps, 14cm, Straight, German
SDP
Catalog No. 50822590
Encompass
Kelly hemostatic forceps are used to clamp blood vessels and hold heavy tissues in place. Kelly forceps, both straight and curved, are available. Our forceps feature a locking ratchet and wider finger rings for easy handling.
Catalog No. 50-822-590 Supplier World Precision Instrument Supplier No. 501241G
Kelly hemostatic forceps are used to clamp blood vessels and hold heavy tissues in place. Kelly forceps, both straight and curved, are available. Our forceps feature a locking ratchet and wider finger rings for easy handling.