Learn More
Med Vet International Miltex Kelly Hemostatic Forceps, 5.5in, Curved
Catalog No. 502829341
Encompass_Preferred
The Miltex 7-38 Kelly forceps are made of OR grade German stainless steel and measure 5". They have a finger ring handle with a ratchet lock and curved tips with serrations.
Catalog No. 50-282-9341 Supplier Med Vet International Supplier No. MIL738
The Miltex 7-38 Kelly forceps are made of OR grade German stainless steel and measure 5". They have a finger ring handle with a ratchet lock and curved tips with serrations.