Learn More
Med Vet International Miltex Kelly Hemostatic Forceps, 5.5in., Straight
Catalog No. 502830532
Encompass_Preferred
The Miltex 7-36 Kelly forceps are made of OR grade German stainless steel and measure 5". They have a finger ring handle with a ratchet lock and straight tips with serrations.
Catalog No. 50-283-0532 Supplier Med Vet International Supplier No. MIL736
The Miltex 7-36 Kelly forceps are made of OR grade German stainless steel and measure 5". They have a finger ring handle with a ratchet lock and straight tips with serrations.