Learn More
Med Vet International Magill Catheter Introducing Forceps, 7.5", Child, German Grade, Von Klaus
Catalog No. 503046814
Encompass_Preferred
7.5in. Magill Catheter Introducing forceps for children is a German grade surgical instrument. They are used to grasp endotracheal tubes during intubation and remvoing foreigh bodies from the airway. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6814 Supplier Med Vet International Supplier No. VK1457502
7.5in. Magill Catheter Introducing forceps for children is a German grade surgical instrument. They are used to grasp endotracheal tubes during intubation and remvoing foreigh bodies from the airway. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.