Learn More
World Precision Instrument Micro Adson Forceps, Straight, German, 12cm
SDP
Catalog No. 50822596
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-822-596 Supplier World Precision Instrument Supplier No. 501244G
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.