Learn More
World Precision Instrument Micro Adson Forceps, 12 cm, 1 x 2 Teeth
SDP
Catalog No. 50822595
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-822-595 Supplier World Precision Instrument Supplier No. 501244
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.