Learn More
World Precision Instrument Microdissecting Forceps, 10.2 cm, Angled, Serrated
SDP
Supplier:  World Precision Instrument 504479
Encompass
WPI Microdissecting Forceps have fine tips for precision work in microsurgery, microdissection, and applications requiring fine manipulation of tissue or small objects under a microscope. The tips on WPI Microdissecting Forceps are only 0.8 mm wide, and you can get straight or angled tips. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. These are made of surgical grade stainless steel, which is suitable for autoclaving or chemical sterilization. Always clean your surgical forceps after use to prevent staining and increase their useable life.
Catalog No. NC0482629
 Restricted Item