Learn More
Roboz Surgical FORCEPS MICRO DISSECTING
SDP
Catalog No. NC9914205
Encompass
Foerster Forceps; Curved; Serrated Tips; 0.5mm Tip Width; 4 in. Length The unique characteristic of the Foerster Forceps is the octagonal handle. The unique handle is designed to provide more feel and control at the tip. This handle also provides a b
Catalog No. NC9914205 Supplier Roboz Surgical Supplier No. RS5101
Foerster Forceps; Curved; Serrated Tips; 0.5mm Tip Width; 4 in. Length The unique characteristic of the Foerster Forceps is the octagonal handle. The unique handle is designed to provide more feel and control at the tip. This handle also provides a b