Learn More
Med Vet International Miltex Halsted Mosquito Hemostatic Forceps, 5in., Curved
Catalog No. 502829342
Encompass_Preferred
The Miltex 7-4 Halsted mosquito forceps are made of OR grade German stainless steel and measure 5" in length. They have a finger ring handle with a ratchet lock and curved tips that are serrated.
Catalog No. 50-282-9342 Supplier Med Vet International Supplier No. MIL74
The Miltex 7-4 Halsted mosquito forceps are made of OR grade German stainless steel and measure 5" in length. They have a finger ring handle with a ratchet lock and curved tips that are serrated.