Learn More
Med Vet International Miltex Halsted Mosquito Hemostatic Forceps, 5in., Straight, Extra Delicate
Catalog No. 502829103
Encompass_Preferred
The Miltex 7-8 Halsted mosquito forceps are made of OR grade German stainless steel and measure 5" in length. They have a finger ring handle with a ratchet lock and curved tips that are straight and extra delicate.
Catalog No. 50-282-9103 Supplier Med Vet International Supplier No. MIL78
The Miltex 7-8 Halsted mosquito forceps are made of OR grade German stainless steel and measure 5" in length. They have a finger ring handle with a ratchet lock and curved tips that are straight and extra delicate.