Learn More
Med Vet International Mosquito Forceps, 3.5 in, Curved, Economy
Catalog No. NC2028706
Encompass_Preferred
Mosquito forceps are used in multiple procedures as a hemostat for small blood vessels. The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries.
This item is not returnable. View return policy
Catalog No. NC2028706 Supplier Med Vet International Supplier No. 4501
Mosquito forceps are used in multiple procedures as a hemostat for small blood vessels. The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries.