Learn More
Med Vet International Miltex Halsted Mosquito Hemostatic Forceps, 5in., Straight
Catalog No. 502831979
Encompass_Preferred
The Miltex 7-2 Halsted mosquito forceps are made of OR grade German stainless steel and measure 5" in length. They have a finger ring handle with a ratchet lock and straight tips that are serrated.
Catalog No. 50-283-1979 Supplier Med Vet International Supplier No. MIL72
The Miltex 7-2 Halsted mosquito forceps are made of OR grade German stainless steel and measure 5" in length. They have a finger ring handle with a ratchet lock and straight tips that are serrated.