Learn More
World Precision Instrument Hartman Mosquito Hemostatic Forceps, 9 cm, Straight, German
SDP
Catalog No. 50822712
Encompass
Halsted (or Halstead) Mosquito Forceps, also called mosquito hemostatic forceps or simply hemostats, are surgical instruments designed for clamping blood vessels or other small tissues to control bleeding during surgery or to provide a secure grip. They are so named because they resemble the slender legs of a mosquito. Life science researchers like these fine hemostats because of the secure grip and the ability to manipulate them in confined spaces. Mosquito Forceps have long, thin, and straight or curved jaws that taper to a fine tip. The jaws are serrated for a firm grasp with minimal tissue trauma. The handles of mosquito forceps have a ratchet lock to secure your grasp once it is engaged.
Catalog No. 50-822-712 Supplier World Precision Instrument Supplier No. 501705G
Halsted (or Halstead) Mosquito Forceps, also called mosquito hemostatic forceps or simply hemostats, are surgical instruments designed for clamping blood vessels or other small tissues to control bleeding during surgery or to provide a secure grip. They are so named because they resemble the slender legs of a mosquito. Life science researchers like these fine hemostats because of the secure grip and the ability to manipulate them in confined spaces. Mosquito Forceps have long, thin, and straight or curved jaws that taper to a fine tip. The jaws are serrated for a firm grasp with minimal tissue trauma. The handles of mosquito forceps have a ratchet lock to secure your grasp once it is engaged.