Learn More
Fine Science Tools Narrow Pattern Forceps
SDP
Catalog No. NC9839859
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 2 x 1.25mm/ Alloy / Material: Stainless Steel/ Length: 12cm. Lighter spring action than standard pattern for grasping more delicate tissue.
Catalog No. NC9839859 Supplier Fine Science Tools Supplier No. 1100312
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 2 x 1.25mm/ Alloy / Material: Stainless Steel/ Length: 12cm. Lighter spring action than standard pattern for grasping more delicate tissue.