Learn More
Roboz Surgical FORCEPS PATTERN NO7 115 MM
SDP
Catalog No. NC9805618
Encompass
Dumont Tweezers; Pattern #7; .07 X .03mm Tip Size; 115mm Length; Material: Dumostar; Biologie Tips
Catalog No. NC9805618 Supplier Roboz Surgical Supplier No. RS4982
Dumont Tweezers; Pattern #7; .07 X .03mm Tip Size; 115mm Length; Material: Dumostar; Biologie Tips