Learn More
Med Vet International Miltex Vantage Peers Towel Clamp, 5.75in, Non-Perforating
Catalog No. 502522260
Encompass_Preferred
The Miltex Vantage V97-520 Peers Forceps are made of Mid-Grade Stainless steel. They have a length of 5.75in. and are often used for securing drapes, suction, ESU or camera cords to the drapes. This forcep has non-perforating jaws.
Catalog No. 50-252-2260 Supplier Med Vet International Supplier No. MILV97520
The Miltex Vantage V97-520 Peers Forceps are made of Mid-Grade Stainless steel. They have a length of 5.75in. and are often used for securing drapes, suction, ESU or camera cords to the drapes. This forcep has non-perforating jaws.