Learn More
Cole-Parmer PTFE-Coated Steel Forceps, 3-7/8" L, Pointed Tips; 2/Pk
Supplier:  Cole-Parmer 0644000
Encompass
PTFE-coated for heat and chemical resistance. Nonsticking and noncorroding. Forceps measures 4-1/4"L. Autoclavable.
Catalog No. NC9427508