Learn More
Electron Microscopy Sciences Forceps with PTFE Tips, Stainless Steel
SDP
Catalog No. 5029025
Encompass_Preferred
4" (100 mm) overall length, 1 1/16" (38 mm) "PTFE" tips; corrosion resistant. Curved shape with a strong fork. Individually blister packed. Ideal for handling balance weights.
Material: Stainless Steel
Catalog No. 50-290-25 Supplier Electron Microscopy Sciences Supplier No. 72978
4" (100 mm) overall length, 1 1/16" (38 mm) "PTFE" tips; corrosion resistant. Curved shape with a strong fork. Individually blister packed. Ideal for handling balance weights.
Material: Stainless Steel