Learn More
Med Vet International Miltex Rankin Forceps, 6.25in, Straight, German, 7-92
Supplier:  Med Vet International MIL792
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Rankin Forceps. Rankin Forceps are a surgery essential, commonly used to clamp blood vessels, hold tissue for dissection, and clamp tissue prior to cauterization or ligation. They are made from premium-grade German stainless steel. Specifications: 6.25in, Straight, 7-92
Catalog No. 50-282-9104