Learn More
Med Vet International Miltex Tartar Removal Forceps, 5in, 6051
Catalog No. 502830421
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Tartar Removal Forceps. The tartar forceps feature a curved beak to allow for the easy removal of heavy calculus from the tooth surface before scaling the teeth. They are made from premium-grade surgical stainless steel. Specifications: 5in, 6051
Catalog No. 50-283-0421 Supplier Med Vet International Supplier No. MIL6051
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Tartar Removal Forceps. The tartar forceps feature a curved beak to allow for the easy removal of heavy calculus from the tooth surface before scaling the teeth. They are made from premium-grade surgical stainless steel. Specifications: 5in, 6051