Learn More
Med Vet International Miltex Vantage Rochester-Carmalt Forceps, 8in, Curved
Catalog No. 502522266
Encompass_Preferred
The Miltex Vantage V97-182 Rochester-Carmalt Forceps have a length of 8in. and are made of mid-grade stainless steel. They have a curved tip and are a great hemostat for clmaping blood vessels and grasping large tissues. These forceps have a textured blade and scissor like ratchet handle that ensures a strong grip with minimal chance of cutting the tissue.
Catalog No. 50-252-2266 Supplier Med Vet International Supplier No. MILV97182
The Miltex Vantage V97-182 Rochester-Carmalt Forceps have a length of 8in. and are made of mid-grade stainless steel. They have a curved tip and are a great hemostat for clmaping blood vessels and grasping large tissues. These forceps have a textured blade and scissor like ratchet handle that ensures a strong grip with minimal chance of cutting the tissue.