Learn More
Med Vet International Miltex MeisterHand Rochester-Ochsner Forceps, 6.25in, Curved, German, MH7-160
Catalog No. 503046757
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Rochester-Ochsner Forceps. Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels. They are made from premium-grade German stainless steel. Specifications: 6.25in, Curved, MH7-160
Catalog No. 50-304-6757 Supplier Med Vet International Supplier No. MILMH7160
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Rochester-Ochsner Forceps. Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels. They are made from premium-grade German stainless steel. Specifications: 6.25in, Curved, MH7-160