Learn More
World Precision Instrument Rochester-Pean Hemostatic Forceps, 19cm, Curved
SDP
Catalog No. 50822717
Encompass
Rochester-Pean forceps may be used in a variety of surgical procedures for clamping blood vessels or other tissues. The solid grip offers surgeons a strong, secure grip, necessary when controlling bleeding. The serrated, grooves along the jaws are designed for an effective grip on tissue to prevent tissue from slipping out of the forceps during use. These hemostatic forceps are available in a variety of sizes, have a locking handle and are constructed of stainless steel. Our premium line is constructed from German stainless steel.
Catalog No. 50-822-717 Supplier World Precision Instrument Supplier No. 501708
Rochester-Pean forceps may be used in a variety of surgical procedures for clamping blood vessels or other tissues. The solid grip offers surgeons a strong, secure grip, necessary when controlling bleeding. The serrated, grooves along the jaws are designed for an effective grip on tissue to prevent tissue from slipping out of the forceps during use. These hemostatic forceps are available in a variety of sizes, have a locking handle and are constructed of stainless steel. Our premium line is constructed from German stainless steel.