Learn More
Med Vet International Miltex Vantage Rochester-Pean Forceps, 6.25in, Curved
Catalog No. 502522267
Encompass_Preferred
The Miltex Vantage V97-138 Rochester-Pean Forceps have a length of 6.25in. and are made of mid-grade stainless steel. They have a curved tip and are a hemostatic forcep with fully serrated jaws. This surgical forcep is commonly used to occlude or clamp blood vessels to control bleeding during surgery.
Catalog No. 50-252-2267 Supplier Med Vet International Supplier No. MILV97138
The Miltex Vantage V97-138 Rochester-Pean Forceps have a length of 6.25in. and are made of mid-grade stainless steel. They have a curved tip and are a hemostatic forcep with fully serrated jaws. This surgical forcep is commonly used to occlude or clamp blood vessels to control bleeding during surgery.